Learn More
Cayman Chemical 1Stearoyl215 SHETEsngl 100ug
SDP
Catalog No. 502937375
Encompass_Preferred
An internal standard for the quantification of 1-stearoyl-2-15(S)-HETE-sn-glycero-3-PE by GC- or LC-MS
Catalog No. 50-293-7375 Supplier Cayman Chemical Supplier No. 27931100 UG
An internal standard for the quantification of 1-stearoyl-2-15(S)-HETE-sn-glycero-3-PE by GC- or LC-MS