Learn More
Cayman Chemical 1Stearoyl215 SHpETEsng 100ug
SDP
Catalog No. 502933635
Encompass_Preferred
An oxidized phospholipid; produced via oxidation of SAPC by 15-LO; toxic to HUVECs at 100 µM
Catalog No. 50-293-3635 Supplier Cayman Chemical Supplier No. 26531100 UG
An oxidized phospholipid; produced via oxidation of SAPC by 15-LO; toxic to HUVECs at 100 µM