Learn More
Cayman Chemical 1Stearoyl2ArachIdonyld11 100ug
SDP
Catalog No. 502940201
Encompass_Preferred
An internal standard for the quantification of 1-stearoyl-2-arachidonoyl-sn-glycero-3-PE by GC- or LC-MS
Catalog No. 50-294-0201 Supplier Cayman Chemical Supplier No. 27929100 UG
An internal standard for the quantification of 1-stearoyl-2-arachidonoyl-sn-glycero-3-PE by GC- or LC-MS