Learn More
Cayman Chemical 1Stearoyl2ArachIdonyld11 100ug
SDP
Supplier:  Cayman Chemical 27928100 UG
Encompass_Preferred
An internal standard for the quantification of 1-stearoyl-2-arachidonoyl-sn-glycero-3-PC by GC- or LC-MS
Catalog No. 50-293-9196