Learn More
Cayman Chemical 1-Stearoyl-2-Docosahexaenoyl-sn-glycero-3-PC, Purity: ≥95%, CAS: 59403-52-0, 10 mg
SDP
Catalog No. 502936702
Encompass_Preferred
A phospholipid; has been used in the study of lipid membrane organization and dynamics; levels are decreased in the L5 anterior horn in a SOD1G93A transgenic mouse model of ALS at the terminal stage of disease
Catalog No. 50-293-6702 Supplier Cayman Chemical Supplier No. 2559510 MG
A phospholipid; has been used in the study of lipid membrane organization and dynamics; levels are decreased in the L5 anterior horn in a SOD1G93A transgenic mouse model of ALS at the terminal stage of disease