Learn More
Cayman Chemical 1-Stearoyl-2-hydroxy-sn-glycero-3-PC, Purity: ≥98%, CAS: 19420-57-6, 100 mg
SDP
Catalog No. 502970103
Encompass_Preferred
A phosphatidylcholine; increases IL-1β production in isolated human blood monocytes at 12.5 µM; plasma levels are decreased in patients with lung cancer
Catalog No. 50-297-0103 Supplier Cayman Chemical Supplier No. 15107100 MG
A phosphatidylcholine; increases IL-1β production in isolated human blood monocytes at 12.5 µM; plasma levels are decreased in patients with lung cancer