Learn More
Cayman Chemical 1Stearoyl2Lauroylracglyc 10mg
SDP
Catalog No. 502932897
Encompass_Preferred
A diacylglycerol; contains stearic acid at the sn-1 position and lauric acid at the sn-2 position; a substrate for the rat diacylglycerol lipase DDHD2
Catalog No. 50-293-2897 Supplier Cayman Chemical Supplier No. 2688310 MG
A diacylglycerol; contains stearic acid at the sn-1 position and lauric acid at the sn-2 position; a substrate for the rat diacylglycerol lipase DDHD2