Learn More
Cayman Chemical 1Stearoyl2LInoleoyl3Oleo 10mg
SDP
Catalog No. 502938953
Encompass_Preferred
A triacylglycerol; has been found in poppy seed, maize, and evening primrose oils
Catalog No. 50-293-8953 Supplier Cayman Chemical Supplier No. 2856210 MG
A triacylglycerol; has been found in poppy seed, maize, and evening primrose oils