Learn More
Cayman Chemical 1Stearoyl2MyrIstoylsngly 100mg
SDP
Catalog No. 502970886
Encompass_Preferred
An asymmetric phospholipid that can interdigitate with equal length (symmetric) acyl chain phospholipids in bilayer membranes to potentially minimize the hydrophobic mismatch between acyl chains
Catalog No. 50-297-0886 Supplier Cayman Chemical Supplier No. 15103100 MG
An asymmetric phospholipid that can interdigitate with equal length (symmetric) acyl chain phospholipids in bilayer membranes to potentially minimize the hydrophobic mismatch between acyl chains