Learn More
Cayman Chemical 1-Stearoyl-2-Myristoyl-sn-glycero-3-PC, Purity: ≥98%, CAS: 20664-02-2, 50 mg
SDP
Catalog No. 502970753
Encompass_Preferred
An asymmetric phospholipid that can interdigitate with equal length (symmetric) acyl chain phospholipids in bilayer membranes to potentially minimize the hydrophobic mismatch between acyl chains
Catalog No. 50-297-0753 Supplier Cayman Chemical Supplier No. 1510350 MG
An asymmetric phospholipid that can interdigitate with equal length (symmetric) acyl chain phospholipids in bilayer membranes to potentially minimize the hydrophobic mismatch between acyl chains