Learn More
Cayman Chemical 1-Stearoyl-2-Oleoyl-3-Linoleoyl-rac-glycerol, Purity: ≥95%, CAS: 2190-14-9, 50 mg
SDP
Catalog No. 502941331
Encompass_Preferred
A triacylglycerol; has been found in peanut and soybean oils as well as ostrich and emu oils; has been used as a substrate for soybean lipoxygenase in cell-free assays
Catalog No. 50-294-1331 Supplier Cayman Chemical Supplier No. 2696850 MG
A triacylglycerol; has been found in peanut and soybean oils as well as ostrich and emu oils; has been used as a substrate for soybean lipoxygenase in cell-free assays