Learn More
Cayman Chemical 1-Stearoyl-2-Oleoyl-sn-glycero-3-PE, Purity: ≥95%, CAS: 6418-95-7, 10 mg
SDP
Catalog No. 502916297
Encompass_Preferred
A phospholipid; has been used in the generation of LNPs for in vitro delivery of mRNA or nuclear-targeted plasmid DNA
Catalog No. 50-291-6297 Supplier Cayman Chemical Supplier No. 3815310 MG
A phospholipid; has been used in the generation of LNPs for in vitro delivery of mRNA or nuclear-targeted plasmid DNA