Learn More
Cayman Chemical 1Stearoyl2Oleoylsnglycer 50mg
SDP
Catalog No. 502916610
Encompass_Preferred
A phospholipid; has been used in the generation of LNPs for the delivery of plasmid DNA in vitro; liposomes containing 1-stearoyl-2-oleoyl-sn-glycero-3-PC and encapsulating HI-6 increase the cerebral AChE reactivation rate by 10% compared with HI-6 alone in a mouse model of soman-induced brain poisoning; has been found in WPPC
Catalog No. 50-291-6610 Supplier Cayman Chemical Supplier No. 3815250 MG
A phospholipid; has been used in the generation of LNPs for the delivery of plasmid DNA in vitro; liposomes containing 1-stearoyl-2-oleoyl-sn-glycero-3-PC and encapsulating HI-6 increase the cerebral AChE reactivation rate by 10% compared with HI-6 alone in a mouse model of soman-induced brain poisoning; has been found in WPPC