Learn More
Cayman Chemical 1-Stearoyl-2-Palmitoyl-rac-glycerol, Purity: ≥98%, CAS: 14015-55-5, 10 mg
SDP
Catalog No. 502936553
Encompass_Preferred
A diacylglycerol containing stearic acid and palmitic acid; has been used to delay drug release in a hydrophilic medium
Catalog No. 50-293-6553 Supplier Cayman Chemical Supplier No. 2559410 MG
A diacylglycerol containing stearic acid and palmitic acid; has been used to delay drug release in a hydrophilic medium