Learn More
Cayman Chemical 1-Stearoyl-3-Linoleoyl-rac-glycerol, Purity: ≥98%, CAS: 126301-96-0, 5 mg
SDP
Catalog No. 502933721
Encompass_Preferred
A diacylglycerol; levels increase in a time-dependent manner during ripening of dry-cured sausages
Catalog No. 50-293-3721 Supplier Cayman Chemical Supplier No. 268305 MG
A diacylglycerol; levels increase in a time-dependent manner during ripening of dry-cured sausages