Learn More
Cayman Chemical 1-Stearoyl-3-Oleoyl-rac-glycerol, Purity: ≥98%, CAS: 18266-27-8, 10 mg
SDP
Catalog No. 502932910
Encompass_Preferred
A diacylglycerol; Intermittent fasting decreases skeletal muscle and hepatic levels in NZO mice; concentration decreases from 4.59 to 1.88% during the dry-curing process of Iberian ham
Catalog No. 50-293-2910 Supplier Cayman Chemical Supplier No. 2687610 MG
A diacylglycerol; Intermittent fasting decreases skeletal muscle and hepatic levels in NZO mice; concentration decreases from 4.59 to 1.88% during the dry-curing process of Iberian ham