Learn More
Apexbio Technology LLC 1STEP UNIV PROBE 4X MIX-1000mg
SDP
Catalog No. 5000897438
Encompass_Preferred
One-step Universal Probe RT-qPCR 4 Mix is a specialized reagent used in molecular biology research primarily for real-time quantitative polymerase chain reactions (RT-qPCR) This mix originates from advancements in genetic and enzymatic technologies facilitating the detection and quantification of RNA targets It works by employing a universal probe-based system that enhances the specificity and efficiency of amplifying target sequences Known for its high sensitivity this mix operates effectively within the nanomolar to low micromolar concentration range It is commonly utilized in applications including gene expression analysis viral load quantification and biomarker discovery in various cell models and occasionally in animal studies This reagent is integral to high-throughput drug screening processes Experimental concentrations typically depend on the specific research design allowing flexibility for different study requirements
Catalog No. 50-008-97438 Supplier Apexbio Technology LLC Supplier No. K15501000
Add to Cart
Add to Cart
One-step Universal Probe RT-qPCR 4 Mix is a specialized reagent used in molecular biology research primarily for real-time quantitative polymerase chain reactions (RT-qPCR) This mix originates from advancements in genetic and enzymatic technologies facilitating the detection and quantification of RNA targets It works by employing a universal probe-based system that enhances the specificity and efficiency of amplifying target sequences Known for its high sensitivity this mix operates effectively within the nanomolar to low micromolar concentration range It is commonly utilized in applications including gene expression analysis viral load quantification and biomarker discovery in various cell models and occasionally in animal studies This reagent is integral to high-throughput drug screening processes Experimental concentrations typically depend on the specific research design allowing flexibility for different study requirements