Learn More
MSC White-Rodgers Thermostat, 1 Heat/1 Cool, 45-90F, 5-1-1 Programmable, 1 ea
Catalog No. 5000505184
Encompass
This standard digital programmable thermostat features 1 heat and 1 cool setting, offering a temperature range of 45–90°F. It provides 5+1+1 programmability for single stage systems, suitable for various heating and cooling applications.
  • Operates from mV to 30 V
  • Electronic switching for reliable operation
  • Features 1 heat, 1 cool style
  • Temperature range of 45–90°F
  • Standard digital 5+1+1 programmable
  • Single stage functionality
  • Horizontal mount type
Catalog No. 50-005-05184 Supplier MSC Supplier No. 05284963
This standard digital programmable thermostat features 1 heat and 1 cool setting, offering a temperature range of 45–90°F. It provides 5+1+1 programmability for single stage systems, suitable for various heating and cooling applications.
  • Operates from mV to 30 V
  • Electronic switching for reliable operation
  • Features 1 heat, 1 cool style
  • Temperature range of 45–90°F
  • Standard digital 5+1+1 programmable
  • Single stage functionality
  • Horizontal mount type