Learn More
Cayman Chemical 1TrIdecnoylracglycerol 100mg
SDP
Catalog No. 502937287
Encompass_Preferred
A monoacylglycerol; has been used as an internal standard for the quantification of mono- and diacylglycerols in buttermilk
Catalog No. 50-293-7287 Supplier Cayman Chemical Supplier No. 26961100 MG
A monoacylglycerol; has been used as an internal standard for the quantification of mono- and diacylglycerols in buttermilk