Learn More
Ace Glass, Inc. PTFE 1in tube compression fitting to 1in male NPT
SDP
Catalog No. 50977688
Encompass
1 PTFE tube compression fitting to 1 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-688 Supplier Ace Glass, Inc. Supplier No. 1270956
1 PTFE tube compression fitting to 1 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.