Learn More
Ace Glass, Inc. PTFE 1in tube compression fitting to 3/4in male NPT
SDP
Supplier:  Ace Glass, Inc. 1270954
Encompass
1 PTFE tube compression fitting to 3/4 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-983-401