Learn More
Chemglass Life Sciences 1µl Inoculating Loop, Blue, Individually Wrapped, Sterile, 400/Pack
SDP
Catalog No. 501897755
Encompass_Preferred
Loops and needles are made from high quality polypropylene and are flexible so stem can be bent to access small volume tubes and dishes. Color coded for easy identification.PP material, hydrophilicUsed in microbiology experimentsDifferent colors for identificationFlexible handles for easier collection and inoculationSterilized by Ethylene OxideEasy-tear sterile individual packaging
Catalog No. 50-189-7755 Supplier Chemglass Life Sciences Supplier No. CGN2458002
Loops and needles are made from high quality polypropylene and are flexible so stem can be bent to access small volume tubes and dishes. Color coded for easy identification.PP material, hydrophilicUsed in microbiology experimentsDifferent colors for identificationFlexible handles for easier collection and inoculationSterilized by Ethylene OxideEasy-tear sterile individual packaging