Learn More
Bellco Glass, Inc. 1µL Inoculating Loop, Bulk
SDP
Catalog No. 502330277
Encompass_Preferred
DescriptionFlexible for easy access to small volume tubes and dishesColor coded for easy identificationGentle on delicate agar or other surfaces Pack Description20/WrapCase Quantity2000SterileYesNon-pyrogenicYesPrimary ColorWhitePrimary MaterialPolypropyleneWorking Volume1μLGraduatedNoRNase/DNase FreeYes
Catalog No. 50-233-0277 Supplier Bellco Glass, Inc. Supplier No. 229601000
DescriptionFlexible for easy access to small volume tubes and dishesColor coded for easy identificationGentle on delicate agar or other surfaces Pack Description20/WrapCase Quantity2000SterileYesNon-pyrogenicYesPrimary ColorWhitePrimary MaterialPolypropyleneWorking Volume1μLGraduatedNoRNase/DNase FreeYes