Learn More
Bellco Glass, Inc. 1µL Inoculating Loop, Indivi
SDP
Supplier:  Bellco Glass, Inc. 229602000
Encompass_Preferred
DescriptionFlexible for easy access to small volume tubes and dishesColor coded for easy identificationGentle on delicate agar or other surfaces Pack Description1/WrapCase Quantity600SterileYesNon-pyrogenicYesPrimary ColorWhitePrimary MaterialPolypropyleneWorking Volume1μLGraduatedNoRNase/DNase FreeYes
Catalog No. 50-233-0557