Learn More
Thomas Scientific 1æl Inoculating Loop, Individual Pack, White, 218mm, Sterile
SDP
Catalog No. 501461514
Encompass_Preferred
CELLTREAT inoculating loops and needles are made from high quality polypropylene and are available in 3 styles; needle, 1µl loop and 10µl loop. The stems are flexible and can be bent to access small volume tubes and dishes and they are color-coded for easy identification. The surface of the loops and needles are vacuum-gas plasma treated, they are packaged in peel-to-open paper/plastic wrappers and are sterile (gamma irradiated). An inoculation loop, also called a smear loop or micro-streaker, is a tool used by biologists to retrieve inoculum using the phenomenon of surface tension. The loop is used to cultivate microbes by streaking agar in a petri dish or plate for subsequent growth. The size of the loop is consistent assuring accurate and repeatable transfers. CELLTREAT L-Shaped Cell Spreaders are ideal for evenly spreading bacterial cultures and are made from high quality polypropylene. They are packaged sterile (gamma irradiated) in individual or bulk, easy-to-ope
Catalog No. 50-146-1514 Supplier Thomas Scientific Supplier No. 1224R34
CELLTREAT inoculating loops and needles are made from high quality polypropylene and are available in 3 styles; needle, 1µl loop and 10µl loop. The stems are flexible and can be bent to access small volume tubes and dishes and they are color-coded for easy identification. The surface of the loops and needles are vacuum-gas plasma treated, they are packaged in peel-to-open paper/plastic wrappers and are sterile (gamma irradiated). An inoculation loop, also called a smear loop or micro-streaker, is a tool used by biologists to retrieve inoculum using the phenomenon of surface tension. The loop is used to cultivate microbes by streaking agar in a petri dish or plate for subsequent growth. The size of the loop is consistent assuring accurate and repeatable transfers. CELLTREAT L-Shaped Cell Spreaders are ideal for evenly spreading bacterial cultures and are made from high quality polypropylene. They are packaged sterile (gamma irradiated) in individual or bulk, easy-to-ope