Learn More
Brookfield Engineer Labs 1UL MICRO PIPETTES VIALS 100PK
Supplier:  Brookfield Engineer Labs 9900150
Encompass
Description1 uL capillary for Vapor Pro sensor calibration and verification. NIST Traceable.100 count. Used with CT-3100(-L), CT-3500RX, and CT-4200XL.
Catalog No. NC2706600