Learn More
LGC GENOMICS LLC BIOSEARCH TECHNOLOGIES INC 1 µmol & 5 µmol Column Holder, For Oligonucleotide Synthesis, Compatible with MerMade Synthesizers, Each
Catalog No. NC2602236
Encompass
The BIOSEARCH TECHNOLOGIES INC 1 µmol & 5 µmol Column Holder is a vital accessory for oligonucleotide synthesis, designed to efficiently handle both 1 µmol and 5 µmol synthesis columns in a 96-well format. It streamlines the synthesis process by securely holding columns in an organized manner, enhancing throughput and ease of use with compatible MerMade synthesizers, setting it apart from other column holders.
Catalog No. NC2602236 Supplier LGC GENOMICS LLC Supplier No. AX192E1033
The BIOSEARCH TECHNOLOGIES INC 1 µmol & 5 µmol Column Holder is a vital accessory for oligonucleotide synthesis, designed to efficiently handle both 1 µmol and 5 µmol synthesis columns in a 96-well format. It streamlines the synthesis process by securely holding columns in an organized manner, enhancing throughput and ease of use with compatible MerMade synthesizers, setting it apart from other column holders.