Learn More
Medchemexpress LLC HY-115400 5mg Medchemexpress, 1V209 CAS:1062444-54-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1154005MG
Encompass_Preferred
Medchemexpress, HY-115400 5mg 1V209 CAS:1062444-54-5 1V209 (TLR7 agonist T7) is a Toll-like receptor 7 (TLR7) agonist and has anti-tumor effects. 1V209 can be conjugated with various polysaccharides to improve its water solubility, and enhance its efficacy, and maintain low toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9381