Description
- 10X to 30X zoom magnification range
- Built-in trinocular camera port for digital imaging applications
- Energy-efficient LED illumination with five lighting settings
- Cordless operation using rechargeable batteries
- One-touch spring-loaded stage clips and right eyepiece diopter adjustment
- Designed for educational, laboratory, and imaging applications
Specifications
Specifications
| Voltage | 110/220 V |
| Magnification Power | 1X to 3X |
| Objective Types | Zoom Stereo |
| Product Type | Stereo Microscope |
| Head Style | Trinocular |
| Lighting Type | LED, Rechargeable Upper and Lower |
| Eyepiece | WF 10X |