Learn More
Apexbio Technology LLC 1X Amplification Diluent 60ml
SDP
Catalog No. 503107129
Encompass_Preferred
Our Tyramide Signal Amplification (TSA) system can detect low-abundance targets in fluorescent immunocytochemistry (ICC) immunohistochemistry (IHC) and in situ hybridization (ISH) techniques enhancing signal sensitivity up to 100-fold The TSA fluorescence kits utilize horseradish peroxidase (HRP) to directly catalyze the covalent deposition of fluorescent groups near the immobilized enzyme The labeling process is rapid (under 10 min) and can be directly visualized through standard or confocal microscope TSA fluorescence kits can be used in combination with anti-fluorescence protocols TSA technology is suitable for brightfield microscopy applications with enzyme conjugates and appropriate chromogenic substrates This product provides reagents used to prepare tyramide fluorescent working solution in TSA products (e g K1050) and is supplied as a ready-to-use solution
Catalog No. 50-310-7129 Supplier Apexbio Technology LLC Supplier No. H100160
Our Tyramide Signal Amplification (TSA) system can detect low-abundance targets in fluorescent immunocytochemistry (ICC) immunohistochemistry (IHC) and in situ hybridization (ISH) techniques enhancing signal sensitivity up to 100-fold The TSA fluorescence kits utilize horseradish peroxidase (HRP) to directly catalyze the covalent deposition of fluorescent groups near the immobilized enzyme The labeling process is rapid (under 10 min) and can be directly visualized through standard or confocal microscope TSA fluorescence kits can be used in combination with anti-fluorescence protocols TSA technology is suitable for brightfield microscopy applications with enzyme conjugates and appropriate chromogenic substrates This product provides reagents used to prepare tyramide fluorescent working solution in TSA products (e g K1050) and is supplied as a ready-to-use solution