Learn More
Quality Biological Inc DPBS without Calcium and Magnesium, 10 Liters
SDP
Catalog No. 50146754
Encompass_Preferred
DPBS is a balanced salt solution (BSS) ¿ Used for the handling and culturing of mammalian cells. ¿ Calcium and magnesium facilitate cell binding and clumping. ¿ DPBS without these ions can be used to wash and rinse suspended cells. ¿ Applications: DPBS serves as an irrigating, transporting or diluting fluid for mammalian cells, while maintaining cell tonicity and viability for a limited period of time. ¿ Used as a diluent of fresh red blood cells to study the trapping of red blood cell aggregate Liters ¿ Wash the wells of microtiter plates ¿ Wash cells during preparation and serial transfer. ¿ Storage Condition: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 12161700 ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-146-754 Supplier Quality Biological Inc Supplier No. 114057151
DPBS is a balanced salt solution (BSS) ¿ Used for the handling and culturing of mammalian cells. ¿ Calcium and magnesium facilitate cell binding and clumping. ¿ DPBS without these ions can be used to wash and rinse suspended cells. ¿ Applications: DPBS serves as an irrigating, transporting or diluting fluid for mammalian cells, while maintaining cell tonicity and viability for a limited period of time. ¿ Used as a diluent of fresh red blood cells to study the trapping of red blood cell aggregate Liters ¿ Wash the wells of microtiter plates ¿ Wash cells during preparation and serial transfer. ¿ Storage Condition: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 12161700 ¿ ¿ Max Shelf Life: 24 months