Learn More
PLANT CELL TECHNOLOGY 1X Dulbecco's Phosphate Buffered Saline Solution without Calcium and Magnesium, Buffer Solution, Mod. 1, Clear Liquid, Sterile, 500mL bottle.
Catalog No. NC3905218
Encompass_Preferred
1X DPBS Mod. 1 is a sterile phosphate-buffered saline solution for mammalian cell culture applications. Provides isotonic environment to maintain cell integrity during washing, resuspension, or media preparation. Supplied as a clear liquid in a 500 mL sterile bottle. Ready-to-use and shelf stable under recommended conditions. Suitable for research, cell culture, and tissue engineering workflows.
Catalog No. NC3905218 Supplier PLANT CELL TECHNOLOGY Supplier No. PBL01500ML
1X DPBS Mod. 1 is a sterile phosphate-buffered saline solution for mammalian cell culture applications. Provides isotonic environment to maintain cell integrity during washing, resuspension, or media preparation. Supplied as a clear liquid in a 500 mL sterile bottle. Ready-to-use and shelf stable under recommended conditions. Suitable for research, cell culture, and tissue engineering workflows.