Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 1X Hank's Balanced Salt Solution

Catalog No. MP091810554 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091810554 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091810554 Supplier MP Biomedicals™ Supplier No. 091810554
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MP Biomedicals™ 1X Hank's Balanced Salt Solution is used to minimize cell attachment or aggregation

  • Does not include calcium, magnesium and phenol red
  • Medium used when growing cells at low densities
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Colorless
Content And Storage Store at 4°C
Concentration 1 X
Product Type Hanks' Balanced Salt Solution
Sterility Sterile
Format Liquid
For Use With (Application) Mammalian Cell Culture
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.