Learn More
Teknova 1X PBS Solution. 1000mL, Sterile, Endotoxin Tested.
SDP
Catalog No. NC9061215
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
This item is not returnable. View return policy
Catalog No. NC9061215 Supplier Teknova Supplier No. P0300
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.