Learn More
Quality Biological Inc PBS pH 7.2, 10 Liters
SDP
Catalog No. 50146752
Encompass_Preferred
PBS (phosphate buffered saline) is a balanced salt solution ¿ Used for a variety biological and cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents ¿ Endotoxin and Mycoplasma tested ¿ Also useful in immunological applications, such as Western Blotting ¿ Storage Condition: 15¿C ¿ 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ pH: 7.1 -7.3 ¿ Osmolality: 300¿5% mOsm/Kg H2O ¿ Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 12161700 ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-146-752 Supplier Quality Biological Inc Supplier No. 114056151
Add to Cart
Add to Cart
PBS (phosphate buffered saline) is a balanced salt solution ¿ Used for a variety biological and cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents ¿ Endotoxin and Mycoplasma tested ¿ Also useful in immunological applications, such as Western Blotting ¿ Storage Condition: 15¿C ¿ 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ pH: 7.1 -7.3 ¿ Osmolality: 300¿5% mOsm/Kg H2O ¿ Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 12161700 ¿ ¿ Max Shelf Life: 24 months