Learn More
Quality Biological Inc PBS pH 7.4 without Calcium and Magnesium, 20 Liters
SDP
Catalog No. 50146757
Encompass_Preferred
PBS (phosphate buffered saline) ¿ Balanced salt solution used for a variety biological and cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. ¿ Endotoxin and Mycoplasma tested ¿ Applications: immunological applications, such as Western Blotting. ¿ Storage Condition: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ pH: 7.3 ¿ 7.5 ¿ Osmolality: 300¿5% mOsm/Kg H2O ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 12161700 ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-146-757 Supplier Quality Biological Inc Supplier No. 114058161
Add to Cart
Add to Cart
PBS (phosphate buffered saline) ¿ Balanced salt solution used for a variety biological and cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. ¿ Endotoxin and Mycoplasma tested ¿ Applications: immunological applications, such as Western Blotting. ¿ Storage Condition: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ pH: 7.3 ¿ 7.5 ¿ Osmolality: 300¿5% mOsm/Kg H2O ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 12161700 ¿ ¿ Max Shelf Life: 24 months