Learn More
Alkali Scientific CellPro™ PBS, 1X, pH 7.4, without Calcium & Magnesium, Sterile Filtered, 500mL
SDP
Catalog No. 502336172
Encompass_Preferred
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.
Catalog No. 50-233-6172 Supplier Alkali Scientific Supplier No. PB500
Add to Cart
Add to Cart
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.