Learn More
Teknova 1X PBS Solution, pH 7.4, 20L Bag, Sterile. 4 Pack.
SDP
Catalog No. 502745892
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
Catalog No. 50-274-5892 Supplier Teknova Supplier No. 213484PK
Add to Cart
Add to Cart
Request a Quote
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.