Learn More
Teknova 1X PBS-TBN Buffer with 0.05% Tween, 1% BSA, 0.1% Sodium Azide. 1000mL, Sterile. 4 pack.
SDP
Catalog No. 502387593
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
Catalog No. 50-238-7593 Supplier Teknova Supplier No. P02204PK
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.