Learn More
Teknova 1X PBS with 4mM EDTA, pH 7.4. 1000mL, Sterile.
SDP
Catalog No. 50842948
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
This item is not returnable. View return policy
Catalog No. 50-842-948 Supplier Teknova Supplier No. P0204
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.