Learn More
Teknova 1X PBS with 0.05% Tween 20, 1% BSA, 1000mL, Sterile.
SDP
Catalog No. 502388169
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
Catalog No. 50-238-8169 Supplier Teknova Supplier No. P0234
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.