Learn More
PLANT CELL TECHNOLOGY 1X Phosphate Buffered Saline Solution, pH 7.4, without Calcium and Magnesium, Buffer Solution, Mod. 6, Clear Liquid, Sterile, 500mL bottle.
Catalog No. NC3905222
Encompass_Preferred
1X PBS Solution Mod. 6, pH 7.4, is a sterile buffer used in mammalian cell culture to maintain physiological pH during washing, dilution, or reagent preparation. Packaged in a clear 500mL bottle. Ready-to-use and stable under recommended conditions. Ideal for research labs, tissue culture, and molecular biology workflows.
Catalog No. NC3905222 Supplier PLANT CELL TECHNOLOGY Supplier No. PBL06500ML
1X PBS Solution Mod. 6, pH 7.4, is a sterile buffer used in mammalian cell culture to maintain physiological pH during washing, dilution, or reagent preparation. Packaged in a clear 500mL bottle. Ready-to-use and stable under recommended conditions. Ideal for research labs, tissue culture, and molecular biology workflows.