Learn More
Bio Rad Laboratories 1X PHOSPHATE BUFFERED SALINE W
Catalog No. 509013079
Encompass
Blocking Reagent; 1X Phosphate Buffered Saline With 1% Casein; Bio-Rad; 10mM sodium phosphate, 150um NaCl, 1% (w/v) casein, pH 7.4; 1L
Catalog No. 50-901-3079 Supplier Bio Rad Laboratories Supplier No. 1610783
Blocking Reagent; 1X Phosphate Buffered Saline With 1% Casein; Bio-Rad; 10mM sodium phosphate, 150um NaCl, 1% (w/v) casein, pH 7.4; 1L