Learn More
Abcam 1 x Red Blood Cell Lysis Buffer, 100ML
Catalog No. 502215682
Encompass_Preferred
Red Blood Cell (RBC) Lysis Buffer ab286858 provides a quick and efficient method of lysing red blood cells. It is provided in a 1x concentrated format, and is also available as ab204733 in a 10x format.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5682 Supplier Abcam Supplier No. AB286858100ML
Red Blood Cell (RBC) Lysis Buffer ab286858 provides a quick and efficient method of lysing red blood cells. It is provided in a 1x concentrated format, and is also available as ab204733 in a 10x format.
The product is subject to the following: Abcam Restricted Use Statement