Learn More
Teknova 1X RPMI-1640 Media without Glucose, Glycine, and Serine. 500mL, Sterile.
SDP
Catalog No. 502388062
Encompass
1X RPMI-1640 without Cerelose Anhydrous Dextrose (Glucose), Glycine, Serine
Catalog No. 50-238-8062 Supplier Teknova Supplier No. R9660
1X RPMI-1640 without Cerelose Anhydrous Dextrose (Glucose), Glycine, Serine