Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1X RPMI 1640 without L-Glutamine and Phosphate, with 0.85 G/L Sodium Bicarbonate, MP Biomedicals

Catalog No. MP091629754 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091629754 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091629754 Supplier MP Biomedicals, Inc Supplier No. 091629754
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • RPMI has successfully been used for the cultivation of normal human and neoplastic leukocytes
  • It is now a popular general purpose medium when properly supplemented
  • As a modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent
  • Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum
  • It has also been widely used in fusion protocols and in the growth of hybrid cells

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00217820
Synonym 1X Liquid

Specifications

Color Red
Quantity 500 mL
Physical Form Solution
Chemical Name or Material 1X RPMI 1640 WITHOUT L-GLUTAMINE AND PHOSPHATE, WITH 0.85 G/L SODIUM BICARBONATE
Recommended Storage Store at +4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.