Learn More
Rockland Immunochemicals 1X SDS-PAGE Destain Solution (Ready-to Use) - MB-022-1000
SDP
Catalog No. 501058986
Encompass_Preferred
Destain Solution is a uniquely formulated, ready-to-use reagent specifically designed for in-gel visualization of Coomassie Blue stained total protein in polyacrylamide gels. The proprietary formulation of the solution ensures high detection sensitivity and low backgrounds.
Catalog No. 50-105-8986 Supplier Rockland Immunochemicals Supplier No. MB0221000
Destain Solution is a uniquely formulated, ready-to-use reagent specifically designed for in-gel visualization of Coomassie Blue stained total protein in polyacrylamide gels. The proprietary formulation of the solution ensures high detection sensitivity and low backgrounds.