Learn More
Rockland Immunochemicals 1X TE pH 8.0 (10 mM Tris HCl, 1.0 mM EDTA) DEPC TREATED - MB-040
SDP
Catalog No. 501058997
Encompass_Preferred
TE buffer is a commonly used buffer solution in molecular biology, especially in procedures involving DNA or RNA. Tris-EDTA buffer is derived from its components: Tris, a common pH buffer, and EDTA, a molecule that chelates cations like Mg2+. The purpose of TE buffer is to solubilize DNA or RNA, while protecting it from degradation.
Catalog No. 50-105-8997 Supplier Rockland Immunochemicals Supplier No. MB040
TE buffer is a commonly used buffer solution in molecular biology, especially in procedures involving DNA or RNA. Tris-EDTA buffer is derived from its components: Tris, a common pH buffer, and EDTA, a molecule that chelates cations like Mg2+. The purpose of TE buffer is to solubilize DNA or RNA, while protecting it from degradation.