Learn More
Rockland Immunochemicals 1X TES pH 7.8 (10 mM Tris HCl, 100 mM NaCl, 1 mM EDTA) DEPC TREATED - MB-058
SDP
Catalog No. 501059016
Encompass_Preferred
1X TES pH 7.8 DEPC TREATED - TES buffer is a commonly used buffer solution in molecular biology, especially in procedures involving DNA or RNA. TES buffer is useful to stabilize proteins, DNA, or RNA, while protecting it from degradation.
Catalog No. 50-105-9016 Supplier Rockland Immunochemicals Supplier No. MB058
1X TES pH 7.8 DEPC TREATED - TES buffer is a commonly used buffer solution in molecular biology, especially in procedures involving DNA or RNA. TES buffer is useful to stabilize proteins, DNA, or RNA, while protecting it from degradation.