Learn More
Bioworld Tris/Glycine Buffer for Resolve Gels (1X), 40 PK(s)
Catalog No. 50198499
Encompass_Preferred
1X Tris/Glycine Buffer for Resolve GelsBuffer System: Pre-weighed powder mix in pouches (packs)1 pouch(pack)= 500 mL
Catalog No. 50-198-499 Supplier Bioworld Supplier No. 208400532
Add to Cart
Add to Cart
1X Tris/Glycine Buffer for Resolve GelsBuffer System: Pre-weighed powder mix in pouches (packs)1 pouch(pack)= 500 mL